Learn More
Hellma™ Quartz Suprasil™ Absorption Cells
Click to view available options
Capacity (Metric):
3.5 mL
Unit Size:
Each
Specifications
Specifications
| Type | Absorption Cell |
| Capacity (Metric) | 3.5 mL |
| Material | Quartz Suprasil |
| Width (Metric) Interior | 9.5mm |
| Quantity | 1Each |
| Path Length | 10 mm |
| Wavelength Range | 200 to 2500 nm |
| Height (Metric) | 49.5 mm |
| Width (Metric) | 12.5 mm |
| For Use With (Application) | For Magnetic Stirrer |