Learn More
Hellma™ Quartz Suprasil Fluorescence Cells
Product Code. 10761493
Click to view available options
Height (Metric):
40 mm
45 mm
49.5 mm
56 mm
Unit Size:
Each
Pack of 2
Product Code. 10761493
Brand: Hellma™ 119004F1040
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Type Fluorescence Cell
Capacity (Metric) 1500 μL
Material Quartz Suprasil
Width (Metric) Interior 4mm
Quantity 2/Pack
Path Length 10 x 4 mm
Height (Metric) 49.5 mm
Width (Metric) 12.5 mm
For Use With (Application) Magnetic Stirrers