Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
Product Code. 11756214
Click to view available options
Capacity (Metric):
0.7 to 5 μL
100 μL
300 μL
Unit Size:
Each
Pack of 2
Product Code. 11756214
Brand: Hellma™ 1052001540
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Type Ultra-micro Absorption Cell
Capacity (Metric) 300 μL
Material Quartz Suprasil
Width (Metric) Interior 2mm
Quantity 2/Pack
Path Length 10 mm
Height (Metric) 25 mm
Width (Metric) 12.5 mm