Learn More
Hellma™ Quartz Suprasils Macro Absorption Cells
Product Code. 15198444
Click to view available options
Capacity (Metric):
26 μL
35,000 μL
Unit Size:
Each
Product Code. 15198444
Brand: Hellma™ 100240
Only null left
Add to basket
Edge
Add to basket
Specifications
Type Macro Absorption Cell
Capacity (Metric) 26 μL
Material Quartz Suprasil
Width (Metric) Interior 9.5mm
Quantity 1 Each
Path Length 2 mm
Height (Metric) 45 mm
Width (Metric) 12.5 mm