Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Click to view available options
For Use With (Equipment):
19/26 socket size
34/35 socket size
40/38 socket size
Unit Size:
Pack of 5
Description
- Support is passed through socket of column and rests at the constriction above the cone
- Manufactured from chemically resistant borosilicate glass
Specifications
Specifications
| For Use With (Application) | Designed to support Raschig rings within plain fractionating columns |
| Type | Column packing support |
| Material | Pyrex Borosilicate Glass |
| For Use With (Equipment) | 34/35 socket size |