Learn More
Quickfit™ Medium Neck Flat Bottom Flask
Useful in reducing problems associated with splashing and frothing.
Brand: Quickfit FF150/3M
Additional Details : Weight : 0.21000kg
Description
- Made of Borosilicate glass
- With longer neck than standard Quickfit flasks
Specifications
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Medium |
150 mL | |
140 mm |