Learn More
Quickfit™ Chromatography Column with Ground Cone
Manufactured from borosilicate glass. Quickfit™ Chromatography Column with Ground Cone includes a 0 porosity chemically resistant glass sintered disc. Upper socket can be connected to a solvent reservoir. Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow.
410.00€ - 498.00€
Specifications
Description | Chromatography Column |
---|---|
Diameter (Metric) Inner | 18 mm |
Quantity | 1 Pack |
Material | Pyrex™ Borosilicate Glass |
Connector Types | 19/26 Cone, 24/29 Socket |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15537770
|
Quickfit
CR32/20 |
310 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15557770
|
Quickfit
CR32/40 |
515 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- With glass sintered disc - porosity 0 to support column packing
- Upper socket can be connected to a solvent reservoir
- Lower cone can be connected to stopcock adapter CR3T/23 to control eluent flow
- Manufactured from borosilicate glass
Specifications
Chromatography Column | |
1 Pack | |
19/26 Cone, 24/29 Socket |
18 mm | |
Pyrex™ Borosilicate Glass | |
Glass sintered disc |