missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Chromatography Column with Rotaflo™ Stopcock

Manufactured from Pyrex™ borosilicate glass. Quickfit™ Chromatography Column with Rotaflo™ Stopcock includes a 0 porosity chemically resistant glass sintered disc. PTFE Stopcock provides fine control of eluent flow. Ground glass socket accepts Quickfit™ reservoirs.

Brand:  Quickfit CR10/10

Additional Details : Weight : 0.06000kg

Product Code. 12468826

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • With PTFE Rotaflo™ Stopcock for fine control of eluent flow
  • Chemically resistant glass sintered disc - porosity 0
  • Ground glass socket accepts Quickfit™ reservoirs
  • Manufactured from Pyrex™ borosilicate glass
Specifications

Specifications

10 mm
1 Pack
Pyrex™ Borosilicate Glass
GP Rotaflo™ 3mm bore stopcock
Chromatography Column
100 mm
14/23 Socket
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions