Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Product Code. 13160063
Change view
Click to view available options
Standard Taper Joint:
10/19
14/23
45/40
NS 12/21
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 7/16
Unit Size:
Pack of 10
Pack of 5
Product Code. Standard Taper Joint unitSize
13160063 14/23 Pack of 10
12421788 10/19 Pack of 10
12491788 45/40 Pack of 5
12481788 NS 34/35 Pack of 5
12471788 NS 29/32 Pack of 10
12461788 NS 24/29 Pack of 10
11339093 NS 7/16 Pack of 10
12451788 NS 19/26 Pack of 10
13150063 NS 12/21 Pack of 10
9 options
Product Code. 13160063
Brand: Quickfit TRF693
Only null left
Add to basket
Add to basket
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Volumetric flasks
Standard Taper Joint 14/23
Material Glass
Product Type Hollow Stopper