Learn More
Quickfit™ Plain Short Stem Adapter
Click to view available options
Overall Length (Metric):
65 mm
120 mm
190 mm
200 mm
Cone Size:
10/19
14/23
19/26
24/29
29/32
Unit Size:
Each
Pack of 5
Description
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
- With short or long stem
- Useful for feeding distillate from condensers into receiving vessels
Specifications
Specifications
| Material | Borosilicate Glass |
| Overall Length (Metric) | 200 mm |
| For Use With (Application) | Feeding distillate from condensers into receiving vessels |
| Cone Size | 24/29 |
| Product Type | Receiver Adapter |