Learn More
Quickfit™ Recovery Still Head Adapter
Click to view available options
Cone Size:
14/23 (Flask/Condenser)
19/26 (Flask/Condenser)
24/29 (Flask), 19/26 (Condenser)
24/29 (Flask/Condenser)
29/32 (Flask/Condenser)
Unit Size:
Each
Description
Recover vapor after distillation. QuickFit™glass recovery head provides an enclosed conduit between flasks and condensers.
Provides a simple vapour conduit between flasks and distillation condensers
Specifications
Specifications
| Material | Borosilicate Glass |
| Cone Size | 19/26 (Flask/Condenser) |
| Product Type | Recovery Still Head Adapter |