Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Click to view available options
Thread Size:
13
18
24
Cone Size:
14/23
19/26
24/29
29/32
missing translation for 'unitSize'
Pack of 2
Beschreibung
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
- With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
- Ideal for securing thermometers, gas inlet tubes, etc
Spezifikation
Spezifikation
| Cone Size | 29/32 |
| For Use With (Application) | Ideal for securing thermometers, gas inlet tubes |
| Includes | Plastic cap, Silicone Rubber ring and PTFE washer |
| Material | Borosilicate Glass |
| Product Type | Cone screw thread adapter |
| Thread Size | 24 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur