Learn More
Quickfit™ Tubing Adapter with Side-arm
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
NS 24/29
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
NS 24/29
Unit Size:
Each
Pack of 5
Description
Quickfit™ Tubing Adapter with Side-arm for attachment of rubber hose
- With sidearm for the attachment of rubber hose etc
- Detachable plastic screwthread connector allows flexible tubing to be fitted easily and safely
Specifications
Specifications
| Cone Size | 14/23 |
| Includes | Detachable Plastic screw thread connector |
| Material | Borosilicate Glass |
| Product Type | Side-arm tubing adapter |
| Standard Taper Joint No. | 14/23 |