Learn More
SVL™ Socket/Screw Joint Adapter
Click to view available options
Standard Taper Joint No.:
15
22
30
Unit Size:
Pack of 10
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Socket fits Quickfit™ cones
- Manufactured from temperature and chemical resistant borosilicate glass
Specifications
Specifications
| For Use With (Equipment) | SVL™ and Quickfit™ Glassware |
| Material | Borosilicate Glass |
| Product Type | Socket/screw thread ground joint adapter |
| Standard Taper Joint No. | 22 |