Learn More
Quinine monohydrochloride dihydrate, 99%
Product Code. p-190300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
10733344 5 g Glass bottle 5g
10458720 25 g Glass bottle 25g
10658894 100 g Glass bottle 100g
3 options
Product Code. 10733344
Brand: Thermo Scientific Acros 163720050
Request Bulk or Custom Format
CAS: 6119-47-7 | C20H29ClN2O4 | 396.91 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6119-47-7
Molecular Formula C20H29ClN2O4
Molecular Weight (g/mol) 396.91
MDL Number MFCD00151248
InChI Key MPQKYZPYCSTMEI-VQYSEXPNNA-N
Synonym quinine hcl dihydrate
PubChem CID 124080947
IUPAC Name (R)-[(2S,4S,5S)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;dihydrate;hydrochloride
SMILES O.O.Cl.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
Specifications
Melting Point 115.0°C to 116.0°C
Color White
Flash Point 122°C
Loss on Drying 6% to 10% (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Fieser 07,311
Merck Index 15, 8177
Specific Rotation Condition − 258.00 (20.00°C c=2,0.1M HCL)
Specific Rotation − 258.00
Solubility Information Solubility in water: 62.5g/L (20 c). Other solubilities: soluble in alcohol,glycerol and chloroform,slightly soluble in ether
Formula Weight 396.92
Percent Purity 99%
Physical Form Crystalline Powder or Silky Needles
Chemical Name or Material Quinine monohydrochloride dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Quinine monohydrochloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.