Learn More
Fungilab™ Alpha Series R Model Rotational Viscometer
Click to view available options
Viscosity Range:
100 to 13.000.000 cP
Unit Size:
Each
Description
- Four line LCD display and keyboard controls
- Multiple language option
- Sound alarm for under or over range
- Universal adapter
- Display of maximum measurable limits
- User-enabled calibration.
- AISI 316 stainless steel spindles
Specifications
Specifications
| Product Type | Viscometer |
| Viscosity Range | 100 to 13.000.000 cP |
| Description | 18 variable speed settings |
| Display Type | 4 lines LCD display and keyborad controls |
| Certifications/Compliance | BS: 6075, 5350 ISO: 2555, 1652 ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716 |
| Includes | The unit is supplied complete with standard spidles (4 spindles for L model, and 6 spindles for R and H models), viscometer stand and spindle protector. |
| Accuracy | 100 to 13.000.000cP |
| Precision | ±1% of full scale |
| Repeatability | 0.2% |
| For Use With (Application) | Measuring viscosity |
| Show More |