Learn More
(R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid, 97%, Thermo Scientific™
Catalog Number p-7034323
Click to view available options
Quantity:
250 mg
1 g
5 g
500788-86-3
C15H21NO5
295.335
MFCD03427937
DNXONWHJJNOKEU-GFCCVEGCSA-N
r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, r-boc-3-methoxy-beta-phe-oh, boc-r-3-amino-3-3-methoxy-phenyl-propionic acid, boc-r-3-amino-3-3-methoxyphenyl propionic acid, r-3-boc-amino-3-3-methoxyphenyl propionic acid, 3r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, 3r-3-tert-butoxy carbonylamino-3-3-methoxyphenyl propanoic acid, pubchem24031, boc-d-?-phe 3-ome-oh, r-boc-3-methoxy-, a-phe-oh
2761760
(3R)-3-(3-methoxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)OC
500788-86-3
C15H21NO5
295.335
MFCD03427937
DNXONWHJJNOKEU-GFCCVEGCSA-N
r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, r-boc-3-methoxy-beta-phe-oh, boc-r-3-amino-3-3-methoxy-phenyl-propionic acid, boc-r-3-amino-3-3-methoxyphenyl propionic acid, r-3-boc-amino-3-3-methoxyphenyl propionic acid, 3r-3-tert-butoxycarbonyl amino-3-3-methoxyphenyl propanoic acid, 3r-3-tert-butoxy carbonylamino-3-3-methoxyphenyl propanoic acid, pubchem24031, boc-d-?-phe 3-ome-oh, r-boc-3-methoxy-, a-phe-oh
2761760
(3R)-3-(3-methoxyphenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC(=CC=C1)OC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals, petrochemicals, and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 106°C to 108°C
Quantity 250 mg
Optical Rotation +40° (c=1 in Ethanol)
Formula Weight 295.34
Percent Purity 97%
Chemical Name or Material (R)-3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid