Learn More
R&D Systems™ Human Erythrocyte Lysing Kit
Product Code. 18731924
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731924 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18731924 Supplier R&D Systems™ Supplier No. WL1000
For the removal of erythrocytes (RBC) from splenocyte preparations.
  • Gently lyses erythrocytes
  • Stabilizes leukocytes for analysis by flow cytometry
  • Maintains natural light scattering and fluorescent staining properties of leukocytes
  • Maintains viability of leukocytes for subsequent tissue culture manipulation
TRUSTED_SUSTAINABILITY
Specifications
Includes H-Lyse Buffer Concentrate (10X); Wash Buffer Concentrate (10X); Fixative Concentrate (10X)
Product Type Cell Lysis Kit
Content And Storage Store at 20°C to 25°C
For Use With (Application) Removal of erythrocytes (red blood cells) from whole blood
Sufficient For 250 mL of whole blood
Quantity 1 Kit