Learn More
R&D Systems™ Recombinant Human Erythropoietin/EPO (Tissue Culture Grade)
Click to view available options
Quantity:
500 U
Unit Size:
1 set
Description
The Recombinant Human Erythropoietin (Tissue Culture Grade) is derived from CHO. The Recombinant Human Erythropoietin (Tissue Culture Grade) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | NP_000790.2 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 2056 |
| Molecular Weight (g/mol) | 21 kDa |
| Name | Erythropoietin/EPO |
| Quantity | 500 U |
| Source | Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein Ala28-Arg193 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 |
| Conjugate | Unconjugated |
| Show More |