missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
R&D Systems™ Human LILRB2/CD85d/ILT4 (NP_005865) VersaClone cDNA
Beskrivning
The Human LILRB2/CD85d/ILT4 (NP_005865) VersaClone cDNA clone contains the complete ORF for ILT4/CD85d/LILRB2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ILT4/CD85d/LILRB2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature.
Specifikationer
Specifikationer
| Product Type | cDNA |
| Description | Gene ID (Entrez): 10288 |
| Gene | ILT4/CD85d/LILRB2 |
| Accession Number | NP_005865 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Insert Type | 1810bp |
| Gene ID (Entrez) | 10288 |
| Gene Alias | CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2 subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Visa mer |
For Research Use Only; Not for Use in Humans
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?