Learn More
R&D Systems™ Mouse Erythrocyte Lysing Kit
Description
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
Key Benefits
- Gently lyses erythrocytes
- Stabilizes leukocytes for analysis by flow cytometry
- Maintains natural light scattering and fluorescent staining properties of leukocytes
- Maintains viability of leukocytes for subsequent tissue culture manipulation
Specifications
Specifications
| Includes | M-Lyse Buffer Concentrate (10X); Wash Buffer Concentrate (10X); Fixative Concentrate (10X) |
| Product Type | Cell Lysis Kit |
| Content And Storage | Store at 20°C to 25°C |
| For Use With (Application) | Removal of erythrocytes (red blood cells) from whole blood |
| Sufficient For | 12.5 x 109 splenocytes |
| Quantity | 1 Kit |