missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IL-17 ELISpot Kit
Description
Includes
- Microplate
- Biotinylated Detection Antibody
- Streptavidin conjugated to Alkaline Phosphatase
- Dilution Buffers
- Wash Buffer Concentrate
- BCIP/NBT Chromogen
- Positive Control
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
Product Features:
- Detect and quantitate individual cells secreting
- High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
- No in vitro expansion of cells required
- High-throughput - ELISpot assays use only a small number of primary cells
Specifications
Specifications
| Antigen | IL-17 |
| Cross Reactivity | Mouse |
| Includes | Mouse IL-17/IL-17A Microplate, Biotinylated Detection Antibody, Streptavidin conjugated to Alkaline Phosphatase, Dilution Buffers, Wash Buffer Concentrate, BCIP/NBT Chromogen, Muse IL-17/IL-17A Positive Control |
| Quantity | 1 x 96-well Plate |
| Sample Type | Whole cells |
| Synonym | CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Detection of cytokine secreting cells, quantitative sandwich ELISA |
| Product Type | Mouse IL-17 ELISpot Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?