missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse SP-D Quantikine ELISA Kit

Product Code. 18722454
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18722454 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722454 Brand R&D Systems™ Brand No. MSFPD0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse SP-D Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse SP-D Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse SP-D in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Conjugate HRP
Assay Sensitivity 9.43 pg/mL
Assay Range 62.50 to 4000 pg/mL
For Use With (Application) ELISA
Sample Type Cell Culture Supernates (50uL), Tissue Lysates (50uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL)
Synonym COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.