missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Sulfamidase/SGSH Protein, CF
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to hydrolyze the substrate 4-Nitrocatechol Sulfate (PNCS). The specific activity is >0.25 pmol/min/µg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | NP_061292 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 6448 |
| Molecular Weight (g/mol) | M.W. (Observed): 60 kDa, reducing conditions; M.W. (theoretical): 56 kDa |
| Name | Sulfamidase/SGSH |
| Quantity | 20 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived mouse Sulfamidase/SGSH protein Arg23-Leu502, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction