missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Rat GDNF Protein

Product Code. 18127670
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18127670

Brand: R&D Systems™ 512GF010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ) at 1 μg/mL binds Recombinant Rat GDNF with an apparent Kd <4 nM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07731
For Use With (Application) Bioactivity, Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) M.W. (theoretical): 15 kDa (monomer)
Name GDNF
Quantity 10 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived rat GDNF protein Ser78-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Gdnf
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein GDNF
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.