missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ELFN1 (NP_001122108) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Beskrivelse
The Human ELFN1 (NP_001122108) VersaClone cDNA clone contains the complete ORF for ELFN1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ELFN1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Ordreinfo
Ships at ambient temperature
Tekniske data
Tekniske data
| Gene Alias | PPP1R28, USP17L7 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion