missing translation for 'onlineSavingsMsg'
Learn More

R)-(+)-N-Benzyl-1-phenylethylamine, TRC

Product Code. 30344342
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30344342

Brand: Toronto Research Chemicals TRCB28873510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-(+)-N-Benzyl-1-phenylethylamine
CAS 38235-77-7
Molecular Formula C15 H17 N
Quantity 10 g
Synonym (αR)-α-Methyl-N-(phenylmethyl)benzenemethanamine, (+)-Benzylphenethylamine, (+)-N-Benzyl-1-phenylethylamine, (+)-N-Benzyl-α-phenylethylamine, (R)-(+)-N-Benzyl-α-methylbenzylamine, (R)-Benzyl(α-methylbenzyl)amine, (R)-N-(α-Methylbenzyl)-N-benzylamine, (R)-N-Benzyl-1-phenyl-1-ethylamine, (R)-N-Benzyl-1-phenylethanamine, (R)-N-Benzyl-1-phenylethylamine, (R)-N-Benzyl-N-α-methylbenzylamine, (R)-N-Benzyl-α-methylbenzylamine, (R)-N-Benzyl-α-methylbenzylamine, Benzyl((R)-1-phenylethyl)amine, N-Benzyl((R)-1-phenylethyl)amine, N-Benzyl-(R)-α-methylbenzylamine, N-[(R)-1-Phenylethyl]benzylamine
InChI Formula InChI=1S/C15H17N/c1-13(15-10-6-3-7-11-15)16-12-14-8-4-2-5-9-14/h2-11,13,16H,12H2,1H3/t13-/m1/s1
SMILES C[C@@H](NCc1ccccc1)c2ccccc2
IUPAC Name (1R)-N-benzyl-1-phenylethanamine
Molecular Weight (g/mol) 211.3
Formula Weight 211.1361
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.