Learn More
(R)-(+)-Styrene oxide, 98%, Thermo Scientific Chemicals
Catalog Number p-7025640
Click to view available options
Quantity:
1 g
5 g
25 g
20780-53-4
C8H8O
120.15
MFCD00066210
AWMVMTVKBNGEAK-QMMMGPOBSA-N
r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
114705
CHEBI:45389
(2R)-2-phenyloxirane
C1O[C@@H]1C1=CC=CC=C1
20780-53-4
C8H8O
120.15
MFCD00066210
AWMVMTVKBNGEAK-QMMMGPOBSA-N
r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r
114705
CHEBI:45389
(2R)-2-phenyloxirane
C1O[C@@H]1C1=CC=CC=C1
It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine. It is the main metabolite of styrene, catalyzed by epoxide hydrolase.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Boiling Point 194°C
Flash Point 74°C (165°F)
Quantity 1 g
UN Number UN2810
Beilstein 1984
Optical Rotation +32° (Neat)
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material (R)-(+)-Styrene oxide