missing translation for 'onlineSavingsMsg'
Learn More

Melford R2B Broth

Product Code. 30018758
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018758

Brand: Melford R24250500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Low nutrient medium formulated without Agar for detecting heterotropic bacteria in water.

Low nutrient medium formulated without Agar for detecting heterotropic bacteria in water. Ideal for membrane filtration methods.

Formula per Liter:

Proteose Peptone 3: 0.5g

Magnesium Sulfate Heptahydrate: 0.1g

Sodium Pyruvate: 0.3g

Casein Acid Hydrolysate: 0.5g

Dipotassium Phosphate: 0.3g

Yeast Extract: 0.5g

Soluble Starch: 0.5g

Glucose: 0.5g

Specifications

Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.