Learn More
Saint-Gobain R3603 Vacuum Tubing
Click to view available options
Thickness (Metric):
1.5 mm
Unit Size:
15m
Description
Crystal clear to allow viewing for possible pump oil back-up which could cause pump damage. Extra heavy walls will withstand full vacuum of 759mmHg at 23°C and 686mmHg at 60°C. Will not swell closed should vacuum pump oil back up. Low vapour pressure makes it suitable for vacuum applications down to 10mmHg to 17mmHg at 21°C. Ideal for analytical testing of gases and vapours, it is also well suited for use in making manometers.
Specifications
Specifications
| Material | Tygon |
| Overall Length (Metric) | 15 m |
| Thickness (Metric) | 1.5 mm |
| Packaging Quantity | 1 Pack |
| Vacuum Level | 759 mm Hg at 80°C |