Learn More
Epredia™ RA Lamb Biopsy Inserts, Biopsy insert pad, white
Prevent the loss of small or friable material while allowing interchange of fluids with Epredia™ RA Lamb Biopsy Inserts.
Brand: Epredia™ E10/87/B
Additional Details : Weight : 0.15000kg
Description
- Mesh fits inside processing cassettes
- Mesh hole size is 0.5mm
Specifications
Insert Pad | |
Stainless Steel | |
Blue |