Learn More
Abnova™ RABAC1 Recombinant Protein
Product Code. p-3721245
Click to view available options
Quantity:
10 μg
25 μg
missing translation for 'unitSize'
10µg
25µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 16185332
missing translation for 'mfr': Abnova™ H00010567P01.10ug
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
Accession Number NP_006414
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 10567
Molecular Weight (g/mol) 45.98
Name RABAC1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MAAQKDQQKDAEAEGLSGTTLLPKLIPSGAGREWLERRRATIRPWSTFVDQQRFSRPRNLGELCQRLVRNVEYYQSNYVFVFLGLILYCVVTSPMLLVALAVFFGACYILYLRTLESKLVLFGREVSPEHQYALAGGISFPFFWLAGAGSAVFWVLGATLVVIGSHAAFHQIEAVDGEELQMEPV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias PRA1/PRAF1/YIP3
Common Name RABAC1
Gene Symbol RABAC1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution