missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60669IR
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat IgG whole molecule | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Goat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| DyLight 755 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction