missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgA Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72777PCP
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| PBS | |
| Human IgA alpha heavy chain | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| PerCP | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction