missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
138.00€ - 228.00€
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5mg/mL |
| Dilution | Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 |
| Applications | Flow Cytometry, Immunomicroscopy |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204563
|
Novus Biologicals
NBP2-60654 |
2 mg |
228.00€
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611589
|
Novus Biologicals
NBP2-60654-0.5mg |
0.5 mg |
138.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.Specifications
| IgG (H+L) | |
| Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
| Polyclonal | |
| Rabbit | |
| IgG | |
| Affinity purified |
| 4.5mg/mL | |
| Flow Cytometry, Immunomicroscopy | |
| Unconjugated | |
| Adaptive Immunity, Immunology | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against human serum proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title