missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Secondary Antibody (20), Novus Biologicals™
Rabbit Monoclonal Antibody
201.00€ - 409.00€
Specifications
| Antigen | IgG1 |
|---|---|
| Clone | 20 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30281469
|
Novus Biologicals
NBP3-43768-20ul |
20 μL |
201.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30281468
|
Novus Biologicals
NBP3-43768-100ul |
100 μL |
409.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISASpecifications
| IgG1 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| Recombinant Mouse IgG1 Fc Protein | |
| Secondary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 20 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title