missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgM (H+L) Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73885-10MG
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgM(H+L) | |
| Polyclonal | |
| HRP | |
| 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 0.01% Gentamicin Sulfate | |
| Mouse IgM whole molecule | |
| 10 mg | |
| This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase and anti-Rabbit Serum. This reagent primarily recognizes Mouse IgM however some light chain cross reactivity will occur against other mouse immunoglobulins. | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| LYOPH | |
| Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500, Immunohistochemistry-Paraffin | |
| Rabbit | |
| Ion exchange chromatography | |
| Secondary | |
| Reconstitute with 0.1mL deionized water. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction