missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-75413
212.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
30 mM Triethanolamine, pH 7.2, 5 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 1 % (w/v) BSA, Protease/IgG free | |
This Rabbit anti-Rat IgG (H+L) Secondary Antibody [Alkaline Phosphatase] was developed against purified rat IgG (H&L). | |
0.5 mg | |
Adaptive Immunity, Immunology | |
Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin | |
Rabbit | |
Affinity purified | |
RUO | |
Secondary | |
Store at 4°C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20°C. Avoid repeated freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction