missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Sheep IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 594
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ SA510056
This item is not returnable.
View return policy
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to chicken, horse, human, mouse and rat.
Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Sheep IgG (H+L) Cross-Adsorbed | |
| Polyclonal | |
| DyLight 594 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Sheep | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 6.8 to 7.4 | |
| Sheep IgG-heavy and light chain. | |
| 500 μg | |
| Secondary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction