Learn More
Atrow™ Rack
Click to view available options
Array:
10 x 10
2 x 6
3 x 8
4 x 12
4 x 3
4 x 6
5 x 10
5 x 5
8 x 16
Height (Metric):
55 mm
65 mm
80 mm
90 mm
100 mm
25 cm
Length (Metric):
100 mm
103 mm
150 mm
172 mm
176 mm
206 mm
215 mm
270 mm
280 mm
296 mm
Unit Size:
Each
Pack of 30
Specifications
Specifications
| Includes | White Rilsan Coating |
| Product Type | Rack |
| Array | 4 x 12 |
| Holds | Tubes (48) |
| For Use With (Application) | 13 mm dia. Hemolysis Tube |
| Quantity | 1 Each |
| Length (Metric) | 206 mm |
| Height (Metric) | 55 mm |