Learn More
Spectrum Labs™ Rack Set for CF-2 Fraction Collector
Click to view available options
For Use With (Application):
Holds 116 x 17 mm or 18 mm diameter tubes
Holds 42 x 28 mm diameter scintilation vials
Unit Size:
Each
Description
- Designed for use with the versatile CF-2 Fraction Collector
- Complete set of tube racks designed to accommodate the tubes chosen
Specifications
Specifications
| Type | Fraction Collector Tube Rack |
| For Use With (Application) | Holds 42 x 28 mm diameter scintilation vials |
| For Use With | CF-2 Fraction Collector |