Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Brand: Hamilton™ 68008-22
Additional Details : Weight : 0.20000kg
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Available pre-sterilized or non-sterile
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifications
200 μL | |
Sterile | |
Virgin Polypropylene |
Non-Filtered | |
10 racks; 960 total tips |