missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein

Human RAF1 (cRAF) [Y340D, Y341D], GST Tag Recombinant Protein

Brand:  Invitrogen™ PV4046

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10211073

  • 7890.00€ / 1mg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RAF1 (cRAF) is a MAP kinase kinase kinase (MAP3K) which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated RAF1 can phosphorylate and activate the dual specificity protein kinases MEK1 and MEK2 which in turn phosphorylates and activates the serine⁄threonine specific protein kinases ERK1 and ERK2. The RAF1 Y340D Y341D mutant is constitutively active.
Specifications

Specifications

NP_002871, Y340D, Y341D
Protein Assays and Analysis, Kinase Assay
66.4kDa
1mg
For research use only. Not for use in diagnostic procedures
RAF1 (cRAF) Y340D Y341D
RAF1 (cRAF) Y340D Y341D was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μg/mL MAP2K1, inactive (P3093) and trace [32P]-γ-ATP for 10 minutes at 30°C
Amino acids 306-648
220nmoles of phosphate transferred to MAP2K1, inactive (P3093) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.25μg/mL
GST Tag
Frozen
0.31mg/mL
Frozen
RAF1 (cRAF) [Y340D] [Y341D] Recombinant Human Protein
–80°C
1 Each
RAF1 (cRAF) [Y340D] [Y341D] Recombinant Human Protein
RAF
RAF1 (cRAF) [Y340D] [Y341D] Recombinant Human Protein
Human
Baculovirus
80% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.