Learn More
Invitrogen™ RAGE (STK30) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMSTK30X5
Additional Details : Weight : 0.33800kg
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Mouse RAGE (STK30) ELISA quantitates Ms RAGE in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms RAGE. Principle of the method The Mouse RAGE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
Q9WVS4 | |
15 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
26448 | |
<12% | |
Pre-coated 96 well plates, Standard, Assay Diluent Concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Other Research | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
13.72 pg/mL to 10,000 pg/mL | |
15 pg/mL to 10,000 pg/mL | |
Biotin | |
Kinase & Inhibitors | |
Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30L, CD30, CD40, CRG-2, CTACK, CXCL16, Eotaxin, Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CSF, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL- | |
Mouse | |
Colorimetric Microplate Reader | |
Mok | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
RAGE1, Rage, Stk30 | |
1 hr. 20 min. |