Learn More
OHAUS™ Ranger™ 4000
Product Code. 15543893 Shop All Ohaus Products
Click to view available options
Capacity:
3.00 kg
6.00 kg
15.00 kg
30.00 kg
Model:
R41ME15
R41ME15-M
R41ME3
R41ME3-M
R41ME30
R41ME30-M
R41ME6
R41ME6-M
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15543893
Brand: OHAUS™ 30237016
Regulatory: This product is under eco-contribution tax.
OHAUS™ Ranger™ 4000 is ideal for industrial weighing applications in which value is important but quality, durability, and functionality cannot be compromised. Equipped with seven application modes scale can provide results for many industrial weighing and measurement needs.
  • Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
  • The most user-friendly scale on the market features Smart Text™ for easy operation and setup to ensure efficiency in the workplace.
  • With the largest display in its class, checkweighing LED indicators and backlit LCD display, the Ranger 4000 can function in almost any industrial environment.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 100 Hours with Rechargeable Battery
Connection Type Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately)
Construction Rugged Cast Aluminum Housing, Stainless Steel Platform
Capacity 3.00 kg
Display Type LED, 3 Color Checkweighing LEDs
Readability 1.000000 g
Linearity 0.200000 g
Model R41ME3-M
Repeatability 0.200000 g
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram
Dimensions (L x W x H) 327 x 315 x 122 mm
Weight (Metric) 7.000 kg