Learn More
Buerkle™ Rapid Couplings, NW 3.2 mm, Male Connections, Hose Nipple with Hose Screw-Joint
Product Code. 15885242 Shop All Bürkle Products
Click to view available options
Tubing Size:
2 mm dia. x 27 mm
4 mm dia. x 37 mm
Material:
POM
Polypropylene
Description:
With valve
Without valve
Unit Size:
Each
Product Code. 15885242
Brand: Buerkle™ 87521104
Only null left
Add to basket
Add to basket
Rapid Couplings, NW 3.2 mm, Male Connections, Hose Nipple with Hose Screw-Joint – Fast and safe connection for liquids, gases, compressed air and vacuum. Buerkle™ Rapid Couplings are ideal when connecting and separating hoses, machines, apparatus, containers and more.
  • Manufactured from polyoxymethylene or polypropylene
  • Maximum flow possible
  • Easy single-handed operation
  • Secure O-ring guarantees that couplings are leakproof
  • Polyoxymethylene operating pressure at 20°C: up to 10 bar, operating temperature from −40° to +80°C
  • Polypropylene operating pressure at 20°C: up to 8 bar, operating temperature from 0° to 110°C
  • Couplings simply connect to each other, automatically clicking into place and opening the valves
  • When disconnecting, the valves are automatically sealed, no leakage occurs
TRUSTED_SUSTAINABILITY
Specifications
Description With valve
Diameter (Metric) Inner 4 mm
Material POM
Diameter (Metric) Outer 6 mm
Overall Length (Metric) 37 mm
For Use With 4 x 6 mm Hose
For Use With (Application) 4x6mm hose inner x outer diameter
Packaging Quantity 1
Tubing Size 4 mm dia. x 37 mm
Width (Metric) 3.2 mm
Includes Rapid coupling
Product Type Rapid Coupling