missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARA Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
RARA Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Brand: Invitrogen PV4415
Additional Details : Weight : 0.50000kg
Description
Retinoic acid receptor alpha, ligand binding domain (RARA-LBD) is a 59.53 kDa recombinant human protein (amino acids 176-462) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells. RARA-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment studies.Specifications
NP_ 000955 | |
Cell Structure, Pharma and Biopharma, Target and Lead Identification and Validation, Ligand Binding Assay | |
59.5kDa | |
50μg | |
For research use only. Not for use in diagnostic procedures | |
Retinoic Acid Receptor alpha | |
3.5nM RAR alpha, LBD (GST) is the final concentration recommended for the LanthaScreen™ TR-FRET RAR alpha Coactivator Assay Kit (PV4409) | |
RARA Recombinant Human Protein | |
GST Tag | |
95% by SDS-PAGE |
0.44mg/mL | |
5914 | |
RARA Recombinant Human Protein | |
–68 to –85°C | |
1 Each | |
RARA Recombinant Human Protein | |
aa 176 to 462 | |
Human | |
Hormone receptors |
For Research Use Only. Not for use in diagnostic procedures.