Learn More
Invitrogen™ RARB Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoic acid receptor beta, ligand binding domain (RARB-LBD) is a 59.20 kDa recombinant human protein (amino acids 168-448) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Brand:  Invitrogen™ PV4716
Additional Details : Weight : 0.50000kg
Product Code. 10155033
  • 1764.00€ / Each
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
RARB-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.
Specifications
Specifications
Nuclear Receptor Assay
Store in ultra-cold freezer (-68 to -85°C).
Ligand Binding Domain
Human
Hormone Receptors
50 μg
RUO - research use only
RAR-Beta LBD
Recombinant
Product Suggestions