Learn More
Invitrogen™ IL-17A Rat Uncoated ELISA Kit with Plates
Description
Includes
- Capture Antibody: Pre-titrated, purified antibody
- Detection Antibody. Pre-titrated, biotin-conjugated antibody
- Standard. Recombinant cytokine for generating standard curve and calibrating samples
- ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
- Assay Diluent. 5X concentrated
- Detection enzyme. Pre-titrated Avidin-HRP
- Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution
The Rat Interleukin 17A (IL-17A) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat IL-17A. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | Q61453 |
| Assay Range | 2-200 pg/mL |
| Assay Sensitivity | 2 pg/mL |
| Conjugate | Biotin |
| Product Type | ELISA Kit |
| Sample Type | Serum, Supernatant |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 301289 |
| Show More |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.