missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IL-6 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
Unit Size:
Each
Description
Rat IL-6 quantitates rat IL-6 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat IL-6.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Specifications
Specifications
| Accession Number | B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; ChIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; Hybridoma growth factor; Ifnb2; IFN-beta-2; IL6; Il-6; ILg6; ILN; Interferon beta-2; Interleukin; interleukin 6; interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; Interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 |
| Assay Range | 30 to 10,000 pg/mL |
| Assay Sensitivity | 30 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 24498 |
| Gene Symbol | Il6 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction